Orders LinkContactUsLinkComponent
Passive air sampling kits
Passive air sampling kits
Replacement frit filters, 2 µm, —, Stainless steel
Catalog # REST24170
Supplier:   Restek
Passive air sampling kits
Catalog # REST24170
Supplier:  Restek
Passive air sampling kits
Passive air sampling kits
Replacement frit filters, 2 µm, —, Stainless steel
Price
64,00 €
Log in for availability
1 * 3 items
Product Details & Documents
This unique grab sampler minimises connections and leaks and houses a critical orifice in the tee fitting.
  • Different orifice sizes for sampling from four minutes to ten hours
  • Compression sample inlet allows easy connection
  • Canister valve connected to a sample port tubing
  • Stainless steel tee with orifice and vaccum gauge
  • Frit filter for insertion into a compression sample inlet
Specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.
 : Canisters and replacement tees and orifices can be ordered separately.
Specifications
  • Description:
    Replacement frit filters, 2 µm
  • Material:
    Stainless steel
  • Seq:
    901
  • Airflow: