Orders LinkContactUsLinkComponent
Capillary nuts
Capillary nuts
Capillary nut
Catalog # REST22688
Supplier:   Restek
Capillary nuts
Catalog # REST22688
Supplier:  Restek
Capillary nuts
Capillary nuts
Capillary nut
Price
83,00 €
Log in for availability
1 * 2 items
Product Details & Documents
These capillary nuts are ideal for Shimadzu 17A, 2010, and 2014 GCs.
The enhanced capillary nut design eliminates the slot, increasing lifetime and durability.
Specifications
  • Description:
    Capillary nut
  • Seq:
    01