Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Auto SYS single taper drilled uniliner (hole near top) for trace, active samples, high recovery and linearity, 6,2 mm, 4,0 mm, 92,1
Catalog # REST21295
Supplier:   Restek
Inlet liners
Catalog # REST21295
Supplier:  Restek
Inlet liners
Inlet liners

Auto SYS single taper drilled uniliner (hole near top) for trace, active samples, high recovery and linearity, 6,2 mm, 4,0 mm, 92,1

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,2 mm
  • Ø int.:
    4,0 mm
  • Description:
    Auto SYS single taper drilled uniliner (hole near top) for trace, active samples, high recovery and linearity
  • Length:
    92,1
  • Seq:
    101