Capillary column nuts, finger-tight
Finger-tight capillary column nuts, for use with standard 1/16"-type ferrules, Stainless steel
Catalog # REST21041
Supplier: Restek
Jump to:
Product Details & Documents
These finger-tight capillary column nuts made from high-quality stainless steel are an alternative to conventional column nuts. These fittings work best with graphite type ferrules and help prevent over tightening, which can cause graphite material to exude into the inlet.
- Allow wrench-free column installations
- Available for standard (1,58 mm) and compact (Agilent-style) ferrules
These fittings can be installed by hand without a wrench and cool down much faster than conventional fittings, making column and inlet maintenance easier.
Specifications
- Description:Finger-tight capillary column nuts, for use with standard 1/16"-type ferrules
- Material:Stainless steel