Order Entry
Export
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Splitless precision liner with deactivated wool for dirty samples, trace samples, 8,0 mm, 5,0 mm, 105
Catalog # REST21028
Supplier:   Restek
Inlet liners
Catalog # REST21028
Supplier:  Restek
Inlet liners
Inlet liners

Splitless precision liner with deactivated wool for dirty samples, trace samples, 8,0 mm, 5,0 mm, 105

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    8,0 mm
  • Ø int.:
    5,0 mm
  • Description:
    Splitless precision liner with deactivated wool for dirty samples, trace samples
  • Length:
    105
  • Seq:
    113