Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Split/splitless, with wool, universal for most common analyses, 5,0 mm, 3,5 mm, 95 mm
  568-2350
 :   Restek
Inlet liners
  568-2350
 :  Restek
Inlet liners
Inlet liners

Split/splitless, with wool, universal for most common analyses, 5,0 mm, 3,5 mm, 95 mm

 
 
   
1 * 1 items
 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    5,0 mm
  • Ø int.:
    3,5 mm
  • Description:
    Split/splitless, with wool, universal for most common analyses
  • Length:
    95 mm
  • Seq:
    073