Inlet liners
Laminar cup splitter for high MW compounds, 5,0 mm, 3,5 mm, 99
REST20869
: Restek
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
- Ext.Ø:5,0 mm
- Ø int.:3,5 mm
- Description:Laminar cup splitter for high MW compounds
- Length:99
- Seq:078



