Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Splitless for trace samples, 5,0 mm, 3,5 mm, 99
Catalog # REST20864
Supplier:   Restek
Inlet liners
Catalog # REST20864
Supplier:  Restek
Inlet liners
Inlet liners

Splitless for trace samples, 5,0 mm, 3,5 mm, 99

Price
Inquire For Price
Log in for availability
1 * 5 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    5,0 mm
  • Ø int.:
    3,5 mm
  • Description:
    Splitless for trace samples
  • Length:
    99
  • Seq:
    077