Order Entry
Export
Orders LinkContactUsLinkComponent
 

CarboBlack columns (packed)

0,19% picric acid, CarboBlack C, PE/Sigma column configuration, Capillary, 2,10 mm, 1,8 m, 80/100
  REST80459-800
 :   Restek
CarboBlack columns (packed)
  REST80459-800
 :  Restek
CarboBlack columns (packed)
CarboBlack columns (packed)

0,19% picric acid, CarboBlack C, PE/Sigma column configuration, Capillary, 2,10 mm, 1,8 m, 80/100

 
 
   
1 * 1 items
 

Graphitised carbon black offers unique selectivity and very little adsorption for alcohol analyses. Two types of CarboBlack supports are available, CarboBlack B and CarboBlack C. CarboBlack B support, with its higher surface area, can hold up to a 10% loading of a non silicone liquid phase. CarboBlack C support can hold up to a 1% loading of a non silicone liquid phase.

  • CarboBlack packings are treated with KOH or picric acid for basic or acidic compounds
 
  • Column Format:
    Capillary
  • Ø int.:
    2,10 mm
  • Mesh size:
    80/100
  • Description:
    0,19% picric acid, CarboBlack C, PE/Sigma column configuration
  • Length:
    1,8 m
  • Seq:
    19