Order Entry
Export
Orders LinkContactUsLinkComponent
 

CarboBlack columns (packed)

6,6% Carbowax 20M, CarboBlack B, Agilent column configuration, Capillary, 2,10 mm, 1,8 m, 80/120
  REST80451-810
 :   Restek
CarboBlack columns (packed)
  REST80451-810
 :  Restek
CarboBlack columns (packed)
CarboBlack columns (packed)

6,6% Carbowax 20M, CarboBlack B, Agilent column configuration, Capillary, 2,10 mm, 1,8 m, 80/120

 
324,26 €
   
1 * 1 items
 

Graphitised carbon black offers unique selectivity and very little adsorption for alcohol analyses. Two types of CarboBlack supports are available, CarboBlack B and CarboBlack C. CarboBlack B support, with its higher surface area, can hold up to a 10% loading of a non silicone liquid phase. CarboBlack C support can hold up to a 1% loading of a non silicone liquid phase.

  • CarboBlack packings are treated with KOH or picric acid for basic or acidic compounds
 
  • Column Format:
    Capillary
  • Ø int.:
    2,10 mm
  • Mesh size:
    80/120
  • Description:
    6,6% Carbowax 20M, CarboBlack B, Agilent column configuration
  • Length:
    1,8 m
  • Seq:
    43