Order Entry
Export
Orders LinkContactUsLinkComponent
3-(Hydroxymethyl)indole

3-(Hydroxymethyl)indole

Glass bottle for solids, 25 g
Catalog # MOLE15557122-25G
Supplier:   Molekula
3-(Hydroxymethyl)indole
Catalog # MOLE15557122-25G
Supplier:  Molekula
CAS Number:  700-06-1
3-(Hydroxymethyl)indole

Glass bottle for solids, 25 g

CAS Number:  700-06-1
Price
115,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
  • CAS Number:700-06-1
  • Formula:C₉H₉NO
  • MDL Number:MFCD00005632
  • Molecular Weight:147.18 g/mol
  • EINECS:211-836-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications