Order Entry
Export
Orders LinkContactUsLinkComponent
L-Threonine

L-Threonine

Glass bottle for solids, 100 g
Catalog # MOLE29578582-100G
Supplier:   Molekula
L-Threonine
Catalog # MOLE29578582-100G
Supplier:  Molekula
CAS Number:  72-19-5
L-Threonine

Glass bottle for solids, 100 g

CAS Number:  72-19-5
Price
63,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
  • CAS Number:72-19-5
  • Formula:C₄H₉NO₃
  • MDL Number:MFCD00064270
  • Molecular Weight:119.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-774-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications