Order Entry
Export
Orders LinkContactUsLinkComponent
Thymolphthalein

Thymolphthalein

Glass bottle for solids, 100 g
Catalog # MOLE40922033-100G
Supplier:   Molekula
Thymolphthalein
Catalog # MOLE40922033-100G
Supplier:  Molekula
CAS Number:  125-20-2
Thymolphthalein

Glass bottle for solids, 100 g

CAS Number:  125-20-2
Price
150,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:12,09542
  • Melting Pt:253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications