Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
QUERCETIN 1 * 1 g
QUERCETIN 1 * 1 g
Print
QUERCETIN 1 * 1 g
Catalog # MOLE86015335-1G
Supplier:
Molekula
QUERCETIN 1 * 1 g
Catalog # MOLE86015335-1G
Supplier:
Molekula
CAS Number:
Expand image
Return to Top
QUERCETIN 1 * 1 g
Add to cart
CAS Number:
Price
76,00 €
Log in
for availability
Unit of Measure
1 * 1 g
-
+
Add to cart
Return Policy
Product Details & Documents