L(+)-Valine
Glass bottle for solids, 100 g
Catalog # MOLE11273495-100G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: H-Val-OH, Valine, (S)-(+)-Valine
- CAS Number:72-18-4
- Formula:C₅H₁₁NO₂
- MDL Number:MFCD00064220
- Molecular Weight:117.15 g/mol
- Merck Index:13,09458
- Boiling Pt:217 °C (1013 hPa)
- Melting Pt:86 °C
- Density:1.284 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-773-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



