Order Entry
Export
Orders LinkContactUsLinkComponent
 

L(-)-Tryptophan

Glass bottle for solids, 1 kg
Catalog # MOLE13831082-1KG
Supplier:   Molekula
L(-)-Tryptophan
Catalog # MOLE13831082-1KG
Supplier:  Molekula
CAS Number:  73-22-3
L(-)-Tryptophan

Glass bottle for solids, 1 kg

CAS Number:  73-22-3
Price
425,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
  • CAS Number:73-22-3
  • Formula:C₁₁H₁₂N₂O₂
  • MDL Number:MFCD00064340
  • Molecular Weight:204.23 g/mol
  • Merck Index:12,09929
  • Melting Pt:290 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-795-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Cat. No.:
    MOLE13831082-1KG

Specifications