L-Threonine
Glass bottle for solids, 1 kg
Catalog # MOLE29578582-1KG
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
- CAS Number:72-19-5
- Formula:C₄H₉NO₃
- MDL Number:MFCD00064270
- Molecular Weight:119.12 g/mol
- Storage Temperature:Ambient
- EINECS:200-774-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



