Ionomycin calcium salt
Glass bottle for solids, 1 mg
Catalog # MOLE39704482-1MG
Jump to:
Discontinued
Product Details & Documents
- CAS Number:56092-82-1
- Formula:C₄₁H₇₀CaO₉
- MDL Number:MFCD00151183
- Molecular Weight:747.1 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mg



