Order Entry
Export
Orders LinkContactUsLinkComponent
3-Nitro-o-xylene

3-Nitro-o-xylene

Glass bottle for solids, 100 g
Catalog # MOLE88984421-100G
Supplier:   Molekula
3-Nitro-o-xylene
Catalog # MOLE88984421-100G
Supplier:  Molekula
CAS Number:  83-41-0
3-Nitro-o-xylene

Glass bottle for solids, 100 g

CAS Number:  83-41-0
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,2-Dimethyl-3-nitrobenzene, 2,3-Dimethylnitrobenzene, 3-Nitro-o-xylene
  • CAS Number:83-41-0
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00007162
  • Molecular Weight:151.16 g/mol
  • Boiling Pt:245 °C (1013 hPa)
  • Density:1.1402 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-474-3
  • UN:1665
  • ADR:6.1,II
  •  :Density: 1.1402 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications