Volumetric flasks, class A, blue markings, BLAUBRAND®
10000* ml, 45/40, 2,00 ± ml, Narrow mouth, with PE stopper
Catalog # BRND2.37236
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Total order from one batch, supplied with batch certificate
* In addition to DIN.
Certifications: DIN EN ISO 1042.
Documents
Certificates
Search by Lot NumberBrand Volumetric Measurement in the Laboratory
Product Literature
BRND2.37290_TDS_EN
Datasheet
Specifications
- NS:45/40
- Capacity:10000* ml
- Description:Narrow mouth, with PE stopper
- Seq:17
- Tolerance:2,00 ± ml