Volumetric flasks, class A, white markings
10 ml, 14/23, 0,06 ± ml
WITG3.670.00114
Borosilicate glass 3.3, amber, with glass stopper.
- White markings
- Conformity certified (DE-M marking)
- Inclusive batch certificate
: DIN EN ISO 1042
- NS:14/23
- Capacity:10 ml
- Seq:03
- Tolerance:0,06 ± ml