Laboratory notebooks, waterproof
Notebook, Blue, Grid, 218×281 mm, 200
Catalog # HEATHS8610B
Supplier: HEATHROW
Jump to:
Product Details & Documents
For admissibility as court evidence, data on laboratory work, experiment details, and design and development. Use these notebooks to keep your indispensable data. Numbered pages include spaces for title, project number, book number, date, and witness signature.
- The covers are water proof and chemical resistant
- Contains periodic and conversion tables in the back
Specifications
- Paper size:218×281 mm
- Type:Grid
- Colour:Blue
- Description:Notebook
- No. of pages:200



