Orders LinkContactUsLinkComponent
Clips for conical joints
Clips for conical joints
Red, 29
Catalog # ROTH1168.1
Supplier:   Roth Carl
Clips for conical joints
Catalog # ROTH1168.1
Supplier:  Roth Carl
Clips for conical joints
Red, 29
Price
56,10 €
Log in for availability
1 * 12 items
Product Details & Documents
These clips for conical joints are made of POM.
  • Resistant to most chemicals
  • Temperature stable up to 90 °C
Delivery information: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Specifications
  • For NS:
    29
  • Colour:
    Red
  • Seq:
    06