Clips for conical joints
Green, 24
Catalog # ROTH1167.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
These clips for conical joints are made of POM.
- Resistant to most chemicals
- Temperature stable up to 90 °C
: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Specifications
- For NS:24
- Colour:Green
- Seq:05