Orders LinkContactUsLinkComponent
Clips for conical joints
Blue, 19
  ROTH1166.1
 :   Roth Carl
Clips for conical joints
  ROTH1166.1
 :  Roth Carl
Clips for conical joints
Blue, 19
52,40 €
1 * 12 items
These clips for conical joints are made of POM.
  • Resistant to most chemicals
  • Temperature stable up to 90 °C
 : Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
  • For NS:
    19
  • Colour:
    Blue
  • Seq:
    04