Clips for conical joints
Yellow, 14
Catalog # ROTH1165.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
These clips for conical joints are made of POM.
- Resistant to most chemicals
- Temperature stable up to 90 °C
: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Specifications
- For NS:14
- Colour:Yellow
- Seq:03