Orders LinkContactUsLinkComponent
Clips for conical joints
Purple, 12
Catalog # ROTH1164.1
Supplier:   Roth Carl
Clips for conical joints
Catalog # ROTH1164.1
Supplier:  Roth Carl
Clips for conical joints
Purple, 12
Price
47,60 €
Log in for availability
1 * 12 items
Product Details & Documents
These clips for conical joints are made of POM.
  • Resistant to most chemicals
  • Temperature stable up to 90 °C
 : Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Specifications
  • For NS:
    12
  • Colour:
    Purple
  • Seq:
    02