Orders LinkContactUsLinkComponent
Clips for conical joints
Clips for conical joints
Green, 10
Catalog # ROTH1163.1
Supplier:   Roth Carl
Clips for conical joints
Catalog # ROTH1163.1
Supplier:  Roth Carl
Clips for conical joints
Green, 10
Price
44,00 €
Log in for availability
1 * 12 items
Product Details & Documents
These clips for conical joints are made of POM.
  • Resistant to most chemicals
  • Temperature stable up to 90 °C
Delivery information: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Specifications
  • For NS:
    10
  • Colour:
    Green
  • Seq:
    01