Order Entry
Export
Orders LinkContactUsLinkComponent
 

Hydrochloric acid 10%

5 L
Catalog # ROTH0710.2
Irritant
Supplier:   Roth Carl
Hydrochloric acid 10%
Catalog # ROTH0710.2
Supplier:  Roth Carl
CAS Number:  7647-01-0
Hydrochloric acid 10%

5 L

CAS Number:  7647-01-0
Price
45,60 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
Specifications

Specifications

  • Pk:
    5 L

Specifications