Orders LinkContactUsLinkComponent
Funnels with short stem
145 mm, 12 mm, 120 mm
  ROTH0587.1
 :   Roth Carl
Funnels with short stem
  ROTH0587.1
 :  Roth Carl
Funnels with short stem
145 mm, 12 mm, 120 mm
2,50 €
1 * 1 items
Funnels, made of natural coloured HDPE, with smooth surface.
With lifting eye and 4 ventilation ribs (except ROTH0583.1).
 : ROTHA828.1 is delivered with sieve Ø 50 mm, mesh size 0,4 mm.
  • Funnel O-Ø:
    120 mm
  • Length:
    145 mm
  • Stem O-Ø:
    12 mm