4-Methyl-2-pentanone
Glass bottle, 2,5 L
Catalog # ROTH0338.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
- CAS Number:108-10-1
- Formula:(CH₃)₂CHCH₂COCH₃
- MDL Number:MFCD00008938
- Molecular Weight:100.16 g/mol
- Merck Index:13,05227
- Boiling Pt:117.4 °C (1013 hPa)
- Melting Pt:–84 °C
- Density:0.801 g/cm³ (20 °C)
- Flash Pt:16 °C
- Storage Temperature:Ambient
- EINECS:203-550-1
- UN:1245
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



