Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Methyl-2-pentanone

Glass bottle, 1 L
Catalog # ROTH0338.1
Flammable
Irritant
Supplier:   Roth Carl
4-Methyl-2-pentanone
Catalog # ROTH0338.1
Supplier:  Roth Carl
CAS Number:  108-10-1
4-Methyl-2-pentanone
4-Methyl-2-pentanone

Glass bottle, 1 L

CAS Number:  108-10-1
Price
134,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
  • CAS Number:108-10-1
  • Formula:(CH₃)₂CHCH₂COCH₃
  • MDL Number:MFCD00008938
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,05227
  • Boiling Pt:117.4 °C (1013 hPa)
  • Melting Pt:–84 °C
  • Density:0.801 g/cm³ (20 °C)
  • Flash Pt:16 °C
  • Storage Temperature:Ambient
  • EINECS:203-550-1
  • UN:1245
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications