Order Entry
Export
Orders LinkContactUsLinkComponent
Brilliant green

Brilliant green

Glass bottle for solids, 100 g
Catalog # ROTH0324.2
Irritant
Supplier:   Roth Carl
Brilliant green
Catalog # ROTH0324.2
Supplier:  Roth Carl
CAS Number:  633-03-4
Brilliant green

Glass bottle for solids, 100 g

CAS Number:  633-03-4
Price
86,10 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
  • CAS Number:633-03-4
  • Formula:C₂₇H₃₄N₂O₄S
  • MDL Number:MFCD00011880
  • Molecular Weight:482.65 g/mol
  • Storage Temperature:Ambient
  • EINECS:211-190-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications