Auramine O
Glass bottle for solids, 25 g
Catalog # ROTH0319.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow
- CAS Number:2465-27-2
- Formula:C₁₇H₂₂ClN₃
- MDL Number:MFCD00012484
- Molecular Weight:303.84 g/mol
- Storage Temperature:Ambient
- EINECS:219-567-2
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



