Orange G
Glass bottle for solids, 25 g
Catalog # ROTH0318.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10
- CAS Number:1936-15-8
- Formula:C₁₆H₁₀N₂Na₂O₇S₂
- MDL Number:MFCD00012457
- Molecular Weight:452.38 g/mol
- Melting Pt:141 °C
- Density:0.45…0.47 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:217-705-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



