Eosin B
Glass bottle for solids, 10 g
Catalog # ROTH0306.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
- CAS Number:548-24-3
- Formula:C₂₀H₆Br₂N₂Na₂O₉
- MDL Number:MFCD00005041
- Molecular Weight:624.06 g/mol
- Merck Index:12,03638
- Melting Pt:> 310 °C
- Storage Temperature:Ambient
- EINECS:208-943-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



