ADA
Glass bottle for solids, 100 g
Catalog # ROTH0250.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
- CAS Number:26239-55-4
- Formula:H₂NC(O)CH₂N(CH₂COOH)₂
- MDL Number:MFCD00008031
- Molecular Weight:190.16 g/mol
- Merck Index:13,00148
- Melting Pt:217…218 °C
- Storage Temperature:Ambient
- EINECS:247-530-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



