Streptomycin sulphate
Glass bottle for solids, 50 g
Catalog # ROTH0236.3
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:3810-74-0
- Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
- MDL Number:MFCD00037023
- Molecular Weight:1457.4 g/mol
- Merck Index:12,08983
- Melting Pt:250 °C
- Storage Temperature:Refrigerator
- EINECS:223-286-0
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



