Order Entry
Export
Orders LinkContactUsLinkComponent
Xylitol

Xylitol

Glass bottle for solids, 250 g
Catalog # ROTH9711.1
Supplier:   Roth Carl
Xylitol
Catalog # ROTH9711.1
Supplier:  Roth Carl
CAS Number:  87-99-0
Xylitol

Glass bottle for solids, 250 g

CAS Number:  87-99-0
Price
29,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
  • CAS Number:87-99-0
  • Formula:C₅H₁₂O₅
  • MDL Number:MFCD00064292
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,10140
  • Boiling Pt:495 °C (1013 hPa)
  • Melting Pt:94 °C
  • Density:1.52 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-788-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications