Toluene
Glass bottle, 1 L
Catalog # ROTH9558.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methylbenzene, Phenylmethane, FM approval
- CAS Number:108-88-3
- Formula:C₆H₅CH₃
- MDL Number:MFCD00008512
- Molecular Weight:92.14 g/mol
- Merck Index:13,09607
- Boiling Pt:110.6 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:0.867 g/cm³ (20 °C)
- Flash Pt:4 °C
- Storage Temperature:Ambient
- EINECS:203-625-9
- UN:1294
- ADR:3,II
- REACH:05-2114615130-69
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



