Order Entry
Export
Orders LinkContactUsLinkComponent
Silver nitrate

Silver nitrate

50 g
Catalog # ROTH9370.1
Oxidizing
Corrosive
Environment
Supplier:   Roth Carl
Silver nitrate
Catalog # ROTH9370.1
Supplier:  Roth Carl
CAS Number:  7761-88-8
Silver nitrate
Silver nitrate

50 g

CAS Number:  7761-88-8
Price
155,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4.35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Specifications

Specifications

  • Pk:
    50 g

Specifications