Cesium sulphate
Glass bottle for solids, 1 kg
Catalog # ROTH9355.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:10294-54-9
- Formula:Cs₂O₄S
- MDL Number:MFCD00010959
- Molecular Weight:361.87 g/mol
- Merck Index:13,02025
- Boiling Pt:1627 °C (1013 hPa)
- Melting Pt:1010 °C
- Density:4.24 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:233-662-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



