Order Entry
Export
Orders LinkContactUsLinkComponent
Cesium sulphate

Cesium sulphate

Glass bottle for solids, 100 g
Catalog # ROTH9355.1
Supplier:   Roth Carl
Cesium sulphate
Catalog # ROTH9355.1
Supplier:  Roth Carl
CAS Number:  10294-54-9
Cesium sulphate

Glass bottle for solids, 100 g

CAS Number:  10294-54-9
Price
166,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:10294-54-9
  • Formula:Cs₂O₄S
  • MDL Number:MFCD00010959
  • Molecular Weight:361.87 g/mol
  • Merck Index:13,02025
  • Boiling Pt:1627 °C (1013 hPa)
  • Melting Pt:1010 °C
  • Density:4.24 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:233-662-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications