Order Entry
Export
Orders LinkContactUsLinkComponent
Sulphur

Sulphur

25 kg
Catalog # ROTH9304.5
Irritant
Supplier:   Roth Carl
Sulphur
Catalog # ROTH9304.5
Supplier:  Roth Carl
CAS Number:  7704-34-9
Sulphur

25 kg

CAS Number:  7704-34-9
Price
207,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
  • CAS Number:7704-34-9
  • Formula:S
  • MDL Number:MFCD00085316
  • Molecular Weight:32.07 g/mol
  • Merck Index:13,09059
  • Boiling Pt:445 °C (1013 hPa)
  • Melting Pt:113…119 °C
  • Density:2.36 g/cm³ (20 °C)
  • Flash Pt:168 °C
  • Storage Temperature:Ambient
  • EINECS:231-722-6
  • UN:1350
  • ADR:4.1,III
Specifications

Specifications

  • Pk:
    25 kg

Specifications