Sulphur
Glass bottle for solids, 1 kg
Catalog # ROTH9304.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:7704-34-9
- Formula:S
- MDL Number:MFCD00085316
- Molecular Weight:32.07 g/mol
- Merck Index:13,09059
- Boiling Pt:445 °C (1013 hPa)
- Melting Pt:113…119 °C
- Density:2.36 g/cm³ (20 °C)
- Flash Pt:168 °C
- Storage Temperature:Ambient
- EINECS:231-722-6
- UN:1350
- ADR:4.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



