Order Entry
Export
Orders LinkContactUsLinkComponent
Zinc stearate

Zinc stearate

Glass bottle for solids, 1 kg
Catalog # ROTH9295.1
Supplier:   Roth Carl
Zinc stearate
Catalog # ROTH9295.1
Supplier:  Roth Carl
CAS Number:  557-05-1
Zinc stearate
Zinc stearate

Glass bottle for solids, 1 kg

CAS Number:  557-05-1
Price
75,30 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Stearic acid zinc salt, Zinc(II) stearate
  • CAS Number:557-05-1
  • Formula:C₃₆H₇₀O₄Zn
  • MDL Number:MFCD00013031
  • Molecular Weight:632.34 g/mol
  • Merck Index:13,10212
  • Melting Pt:140 °C
  • Density:1.095 g/cm³ (20 °C)
  • Flash Pt:276 °C
  • Storage Temperature:Ambient
  • EINECS:209-151-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications