Hydrochloric acid 37%
Glass bottle, 2,5 L
Catalog # ROTH9277.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46 g/mol
- Boiling Pt:110 °C (1013 hPa)
- Melting Pt:–30 °C
- Density:1.18 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-595-7
- UN:1789
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



