Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium alginate

Sodium alginate

Glass bottle for solids, 1 kg
Catalog # ROTH9180.2
Supplier:   Roth Carl
Sodium alginate
Catalog # ROTH9180.2
Supplier:  Roth Carl
CAS Number:  9005-38-3
Sodium alginate

Glass bottle for solids, 1 kg

CAS Number:  9005-38-3
Price
141,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Alginic acid sodium salt, Algin
  • CAS Number:9005-38-3
  • MDL Number:MFCD00081310
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications