Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium alginate

Sodium alginate

Glass bottle for solids, 250 g
Catalog # ROTH9180.1
Supplier:   Roth Carl
Sodium alginate
Catalog # ROTH9180.1
Supplier:  Roth Carl
CAS Number:  9005-38-3
Sodium alginate

Glass bottle for solids, 250 g

CAS Number:  9005-38-3
Price
49,30 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Alginic acid sodium salt, Algin
  • CAS Number:9005-38-3
  • MDL Number:MFCD00081310
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications