Order Entry
Export
Orders LinkContactUsLinkComponent
PIPES

PIPES

Glass bottle for solids, 500 g
Catalog # ROTH9156.4
Supplier:   Roth Carl
PIPES
Catalog # ROTH9156.4
Supplier:  Roth Carl
CAS Number:  5625-37-6
PIPES
PIPES

Glass bottle for solids, 500 g

CAS Number:  5625-37-6
Price
296,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
  • CAS Number:5625-37-6
  • Formula:C₈H₁₈N₂O₆S₂
  • MDL Number:MFCD00006159
  • Molecular Weight:302.37 g/mol
  • Merck Index:12,07633
  • Boiling Pt:499 °C (1013 hPa)
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:227-057-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications